Thaumatin, Thaumatococcus daniellii
Thermo Scientific Chemicals

Thaumatin, Thaumatococcus daniellii

A mixture of the intensely sweet proteins thaumatin I and thaumatin II | CAS: 53850-34-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
1 g
Catalog number J63668.03
also known as J63668-03
Price (USD)
339.65
Special offer
Online exclusive
Ends: 30-Jun-2026
399.00
Save 59.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
339.65
Special offer
Online exclusive
Ends: 30-Jun-2026
399.00
Save 59.35 (15%)
Each
Specifications
CAS53850-34-3
Chemical Name or MaterialThaumatin
ColorBrown to Yellow
EINECS Number258-822-2
MDL NumberMFCD00132420
View more
Thaumatin is used as a mixture of the intensely sweet proteins, used to enhance the taste of various foods, Flavoring Agents, Food Additives, Sweetening Agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thaumatin is used as a mixture of the intensely sweet proteins, used to enhance the taste of various foods, Flavoring Agents, Food Additives, Sweetening Agents.

Solubility
Soluble in water (25 mg/ml), and 60% ethanol. Insoluble in ether, and benzene .

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Maria Szwacka.; Magdalena Krzymowska.; Anita Osuch.; Magdalena Ewa Kowalczyk.; Stefan Malepszy. Variable properties of transgenic cucumber plants containing the thaumatin II gene from Thaumatococcus daniellii. Acta Physiologiae Plantarum. 2002, 24, (2), 173-185.
  2. van der Wel, H.; van Soest, T.C.; Royers, E.C. Crystallization and crystal data of thaumatin I, a sweet-tasting protein from Thaumatococcus daniellii benth. FEBS Letters. 1975, 56, (2), 316-317.