Isosorbide mononitrate, 98+%
Isosorbide mononitrate, 98+%
Isosorbide mononitrate, 98+%
Isosorbide mononitrate, 98+%
Thermo Scientific Chemicals

Isosorbide mononitrate, 98+%

A coronary vasodilator that does not require hepatic biotransformation | CAS: 16051-77-7 | C6H9NO6 | 191.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63674.09
also known as J63674-09
Price (USD)
322.65
Online Exclusive
358.00
Save 35.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
322.65
Online Exclusive
358.00
Save 35.35 (10%)
Each
Add to cart
Specifications
CAS16051-77-7
ChEBICHEBI:6062
Chemical Name or MaterialIsosorbide mononitrate
ColorWhite
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: ISOSORBIDE-5-MONONITRATE
View more
A coronary vasodilator that does not require hepatic biotransformation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only