Dextran, MW ca 500,000
Dextran, MW ca 500,000
Dextran, MW ca 500,000
Thermo Scientific Chemicals

Dextran, MW ca 500,000

Dextran, CAS # 9004-54-0, is a glucose polymer. It is mainly used to increase electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity). | CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63702.09
also known as J63702-09
Price (USD)
47.30
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
47.30
Each
Chemical Identifiers
CAS9004-54-0
IUPAC NameDextran
Molecular Formula(C6H9O5)n(C6H10O5)m
SMILESO[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
MDL NumberMFCD00130935
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Solution TestColor of solution at A375nm: ≤0.15 OD (6% solution in water)
Loss on Drying≤7.0%
Solution TestCold water solubility: soluble in 1%
Viscosity0.56 to 0.70 at 37°C (1% solution in water)
View more
It is used in increasing the electronegativity and reducing erythrocyte aggregation and platelet adhesiveness and also serves as a plasminogen activator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides

Application

  • Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)
  • This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin
  • Dextran can be used to reduce thrombosis in animal models
  • It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules
  • Dextran can be used as an immobilization agent in biosensors
RUO – Research Use Only