Ketotifen fumarate, 99%
Ketotifen fumarate, 99%
Ketotifen fumarate, 99%
Ketotifen fumarate, 99%
Thermo Scientific Chemicals

Ketotifen fumarate, 99%

CAS: 34580-14-8 | C23H23NO5S | 425.499 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63708.MD
also known as J63708-MD
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.30
Save 9.65 (15%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.30
Save 9.65 (15%)
Each
Add to cart
Chemical Identifiers
CAS34580-14-8
IUPAC Name(2E)-but-2-enedioic acid; 2-(1-methylpiperidin-4-ylidene)-6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-8-one
Molecular FormulaC23H23NO5S
InChI KeyYNQQEYBLVYAWNX-WLHGVMLRSA-N
SMILESOC(=O)C=CC(O)=O.CN1CCC(CC1)=C1C2=C(SC=C2)C(=O)CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.5%
Appearance (Color)White to pale cream to pale yellow
FormPowder
Mass SpecConforms to structure
Ketotifen fumaratin is an H1 receptor antagonist. In its ophthalmic form, it is used to treat allergic conjunctivitis, or the itchy red eyes caused by allergies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (10 mg/ml), DMSO (100 mM), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. MD Gregg J. Berdycorrespondence,; MD Dennis L. Spangler,; MD George Bensch,; MD Susan S. Berdy,; OD Robert C. Brusatti. A comparison of the relative efficacy and clinical performance of olopatadine hydrochloride 0.1% ophthalmic solution and ketotifen fumarate 0.025% ophthalmic solution in the conjunctival antigen challenge model. Clinical therapeutics. 2000, 22 (7), 826-833.
  2. Mayank Joshi,; Ambikanandan Misra.Dry powder inhalation of liposomal Ketotifen fumarate: formulation and characterization. International Journal of Pharmaceutics. 2001, 223 (1-2).