Imipramine hydrochloride
Imipramine hydrochloride
Imipramine hydrochloride
Thermo Scientific Chemicals

Imipramine hydrochloride

A serotonin uptake inhibitor | CAS: 113-52-0 | C19H25ClN2 | 316.873 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63723.06
also known as J63723-06
Price (USD)
74.30
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
74.30
Each
Chemical Identifiers
CAS113-52-0
IUPAC Namehydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
Molecular FormulaC19H25ClN2
InChI KeyXZZXIYZZBJDEEP-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisNitrogen: 8.5 - 9.1%
Thin Layer Chromatography≥ 99%
Elemental AnalysisCarbon: 70.6 - 73.5%
Solution TestTurbidity (50 mg/ml): Clear
Appearance (Color)White to off White Powder
Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Amit Kumar Jain.; Narisetty Sunil Thomas.; Ramesh Panchagnula. Transdermal drug delivery of imipramine hydrochloride.: I. Effect of terpenes. Journal of Controlled Release. 2002, 79, (1-3), 93-101.
  2. Elsa-Grace V. Giardina.; Thomas Bigger Jr, J. Antiarrhythmjc effect of imipramine hydrochloride in patients with ventricular premature complexes without psychological depression ☆. The American Journal of Cardiology. 1982, 50, (1), 172-179.