Difloxacin hydrochloride
Difloxacin hydrochloride
Difloxacin hydrochloride
Thermo Scientific Chemicals

Difloxacin hydrochloride

CAS: 91296-86-5 | C21H20ClF2N3O3 | 435.856 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63754.06
also known as J63754-06
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Specifications
Beilstein6464798
CAS91296-86-5
Chemical Name or MaterialDifloxacin hydrochloride
ColorWhite
Formula Weight435.85
View more
A quinolone antibacterial compound. Structurally related to norfloxacin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A quinolone antibacterial compound
  • Structurally related to norfloxacin
RUO – Research Use Only