Gossypol, 98+%
Gossypol, 98+%
Gossypol, 98+%
Thermo Scientific Chemicals

Gossypol, 98+%

Lipophilic agent derived from cottonseed | CAS: 303-45-7 | C30H30O8 | 518.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
Catalog number J63767.MC
also known as J63767-MC
Price (USD)
193.65
Online exclusive
215.00
Save 21.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
193.65
Online exclusive
215.00
Save 21.35
Each
Chemical Identifiers
CAS303-45-7
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0%
Proton NMRConforms to structure
FormPowder
Gossypol acts as an inhibitor for several dehydrogenase enzymes. Gossypol has also long been known to possess antimalarial properties. Gossypol provided reliable contraception. gossypol provokes infertility and causes spermatogenesis. Gossypol is used as an active antifertility agent in a variety of male animals, including mice, rats, hamsters, monkeys, rabbits, and bulls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gossypol acts as an inhibitor for several dehydrogenase enzymes. Gossypol has also long been known to possess antimalarial properties. Gossypol provided reliable contraception. gossypol provokes infertility and causes spermatogenesis. Gossypol is used as an active antifertility agent in a variety of male animals, including mice, rats, hamsters, monkeys, rabbits, and bulls.

Solubility
Soluble in 100%ethanol (25 mg/ml), DMF (25 mg/ml), acetone, DMSO (25 mM), methanol (2 mg/ml), ether, chloroform, sodium carbonate, and dilute aqueous solutions of ammonia . Insoluble in water. Gossypol is a male antifertility agent with antispermatogenic activity and has been shown to contain antitumor, anitviral, and antioxidant properties.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. M.C. Chang,; Zhiping Gu (Chi-ping Ku),; S.K. Saksena. Effects of Gossypol on the fertility of male rats, hamsters and rabbits. Contraception. 1980, 21(5), 461-469.
  2. Ying-Bo Mao,; Wen-Juan Cai,; Jia-Wei Wang,; Gao-Jie Hong. Silencing a cotton bollworm P450 monooxygenase gene by plant-mediated RNAi impairs larval tolerance of gossypo. Nature Biotechnology. 2007 , 251307 - 1313 .