Acebutolol hydrochloride
Acebutolol hydrochloride
Acebutolol hydrochloride
Thermo Scientific Chemicals

Acebutolol hydrochloride

Cardioselective beta-adrenergic blocker | CAS: 34381-68-5 | C18H29ClN2O4 | 372.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63790.09
also known as J63790-09
Price (USD)
274.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
274.00
Each
Chemical Identifiers
CAS34381-68-5
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥94.0%
Appearance (Color)White to off-white
Cardioselective beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only