Brilliant Blue G soln., Ready-to-Use
Brilliant Blue G soln., Ready-to-Use
Brilliant Blue G soln., Ready-to-Use
Thermo Scientific Chemicals

Brilliant Blue G soln., Ready-to-Use

CAS: 6104-58-1 | C47H48N3NaO7S2 | 854.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J63797.K2
also known as J63797-K2
Price (USD)
175.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
175.00
Each
Chemical Identifiers
CAS6104-58-1
IUPAC Namesodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Molecular FormulaC47H48N3NaO7S2
InChI KeyCMGWKYXJIYYESN-UHFFFAOYSA-M
SMILES[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
ConcentrationCoomassie Blue G: 0.2%
Appearance (Color)Blue
ConcentrationMethanol: 30%
ConcentrationAcetic Acid: 7%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only