CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals
CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals
CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals

5-HT-1B full agonist
Have Questions?
Catalog NumberQuantity
J63801.MA
also known as J63801-MA
10 mg
Catalog number J63801.MA
also known as J63801-MA
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Specifications
CAS109028-10-6
Chemical Name or MaterialCGS 12066B dimaleate
ColorWhite
Formula Weight566.49
IUPAC Name(E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
View more
5-HT-1B full agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only