Toremifene, 98+%
Thermo Scientific Chemicals

Toremifene, 98+%

A chlorinated tamoxifen analogue that induces apoptosis in some cells | CAS: 89778-26-7 | C26H28ClNO, C26H28ClNO | 405.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63803.03
also known as J63803-03
Price (USD)
199.65
Online exclusive
222.00
Save 22.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
199.65
Online exclusive
222.00
Save 22.35
Each
Chemical Identifiers
CAS89778-26-7
IUPAC Name(2-{4-[(1Z)-4-chloro-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
Molecular FormulaC26H28ClNO
InChI KeyXFCLJVABOIYOMF-QPLCGJKRSA-N
SMILESCN(C)CCOC1=CC=C(C=C1)C(=C(\CCCl)C1=CC=CC=C1)\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥98.0%
Appearance (Color)White
Impurity content106.0-110.0°C
FormPowder or crystals or crystalline powder
A chlorinated tamoxifen analogue, that induces apoptosis in some cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only