This product is animal origin and can only be shipped within the United States
Hemoglobin, Bovine
Thermo Scientific Chemicals

Hemoglobin, Bovine

For research use | 64.5KDa
Have Questions?
Change viewbuttonViewtableView
Quantity:
5g
25g
100g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63838.22
also known as J63838-22
Price (USD)
-
Quantity:
100g
Request bulk or custom format
Chemical Identifiers
CAS9008-02-0
MDL NumberMFCD00131282
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to brown
FormPowder or crystalline powder
Loss on Drying≤10.0%
pH7.0-9.0
Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide. It is used as a coloring agent for pet foods, a natural source of iron for nutraceuticals, a protein source for non-ruminant animals, and as a raw material for pharmaceutical porphyrin derivative production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only