This product is animal origin and can only be shipped within the United States
Mucin, Bovine Submaxillary Gland
Thermo Scientific Chemicals

Mucin, Bovine Submaxillary Gland

Mucin, CAS # 84195-52-8, is a high molecular weight glycoprotein, and is a major constituent of saliva, gastrointestinal juices, and other secretions. It is used as a substrate for the viral enzyme neuraminidase. | 400-4000KDa
Have Questions?
Change viewbuttonViewtableView
Quantity:
100mg
500mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63859.ME
also known as J63859-ME
Price (USD)
-
Quantity:
500mg
Request bulk or custom format
Chemical Identifiers
CAS84195-52-8
SpecificationsSpecification SheetSpecification Sheet
CommentSuitable as a neuramindase substrate.
Appearance (Color)White to pale brown
FormPowder
Mucin is used in a study to investigate a galactose-specific lectin from the red marine alga Ptilota filicina. Mucin from bovine submaxillary glands has also been used in a study to investigate the characterization of a unique mucin-like glycoprotein secreted by a human endometrial adenocarcinoma cell line, neuraminidase substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Mucin derived from the bovine submaxillary gland is a glycoprotein present in saliva, gastrointestinal juices, and other secretions that promote cilia lubrication

Application

  • Mucin can be used as a source of O-linked oligosaccharides for the viral enzyme, neuraminidase
  • Mucins are glycoproteins with roles in immunity and digestion
  • In biological research, mucin is used to investigate a galactose-specific lectin from the red marine alga Ptilota filicina
  • It is used to investigate the characterization of a unique mucin-like glycoprotein secreted by a human endometrial adenocarcinoma cell line
  • It is used as a substrate for recombinant sialidase Gardnerella vaginalis
  • It is used in protocols to quantify mucin
RUO – Research Use Only