Carbonate, 0.5M buffer soln., pH 9.0
Carbonate, 0.5M buffer soln., pH 9.0
Carbonate, 0.5M buffer soln., pH 9.0
Thermo Scientific Chemicals

Carbonate, 0.5M buffer soln., pH 9.0

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J63899.AK
also known as J63899-AK
Price (USD)
84.90
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
84.90
Each
Specifications
Concentration0.5 M buffer soln.
Quantity250 mL
TypeCarbonate buffer soln.
pH Range9.0
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only