What is the concentration of Phosphatase Inhibitor Cocktail I (Cat. No. J63907.AA)?
Phosphatase Inhibitor Cocktail I is a 100X concentrated solution with the following component concentrations:
- 200 mM imidazole
- 100 mM sodium fluoride
- 115 mM sodium molybedate dihydrate
- 100 mM sodium orthovanadate
- 400 mM sodium tartrate dihydrate