BICINE, 0.5M buffer soln., pH 8.0
BICINE, 0.5M buffer soln., pH 8.0
BICINE, 0.5M buffer soln., pH 8.0
Thermo Scientific Chemicals

BICINE, 0.5M buffer soln., pH 8.0

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J63924.AE
also known as J63924-AE
Price (USD)
112.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
112.00
Each
Specifications
CAS150-25-4
ChEBICHEBI:40957
Chemical Name or MaterialBICINE buffer soln.
IUPAC Name2-[bis(2-hydroxyethyl)amino]acetic acid
InChI KeyMFBDBXAVPLFMNJ-UHFFFAOYSA-M
View more
BICINE is used for the preparation of stable substrate solution for serum guanase determination. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BICINE is used for the preparation of stable substrate solution for serum guanase determination. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only