Dynorphin A (1-13), Porcine
Thermo Scientific Chemicals

Dynorphin A (1-13), Porcine

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J64015.LB0
also known as J64015-#0
Price (USD)
232.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
232.00
Each
Chemical Identifiers
CAS72957-38-1
SpecificationsSpecification SheetSpecification Sheet
FormLyophilized powder
AssayPeptide content: Report Value
Assay (unspecified)Peptide content: Report Value
AssayPeptide Purity: Report Value
Mass SpecExhibits correct MW
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C