Prima-1, Thermo Scientific Chemicals
Prima-1, Thermo Scientific Chemicals
Prima-1, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Prima-1, Thermo Scientific Chemicals

A cell-permeable quinuclidinone analog
Have Questions?
Catalog NumberQuantity
J64071.MA
also known as J64071-MA
10 mg
Catalog number J64071.MA
also known as J64071-MA
Price (USD)
230.65
Online exclusive
256.00
Save 25.35
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
230.65
Online exclusive
256.00
Save 25.35
Each
Specifications
CAS5608-24-2
Chemical Name or MaterialPrima-1
ColorWhite
Formula Weight185.2
Health Hazard 1H302-H317
View more
A cell-permeable quinuclidinone analog.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only