(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals
(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals
(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals

A NMDA agonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
10 mg
Catalog number J64075.MCR
also known as J64075-M^
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Chemical Identifiers
CAS23052-81-5
IUPAC Name(2S)-2-azaniumyl-4-phosphonatobutanoate
Molecular FormulaC4H8NO5P
InChI KeyDDOQBQRIEWHWBT-VKHMYHEASA-L
SMILES[NH3+][C@@H](CCP([O-])([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical RotationReport value
FormPowder
Purity≥96.0%
A NMDA agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only