Dp44mT
Dp44mT
Dp44mT
Thermo Scientific Chemicals

Dp44mT

CAS: 152095-12-0 | C14H15N5S | 285.369 g/mol
Have Questions?
Catalog NumberQuantity
J64090.MB
also known as J64090-MB
25 mg
Catalog number J64090.MB
also known as J64090-MB
Price (USD)
113.65
Online exclusive
126.00
Save 12.35
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
113.65
Online exclusive
126.00
Save 12.35
Each
Chemical Identifiers
CAS152095-12-0
IUPAC Name1-{[bis(pyridin-2-yl)methylidene]amino}-3,3-dimethylthiourea
Molecular FormulaC14H15N5S
InChI KeyXOBIGRNRXCAMJQ-UHFFFAOYSA-N
SMILESCN(C)C(=S)NN=C(C1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Purity≥95.0%
FormSolid
An iron chelator and selective anticancer agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only