Potassium chloride, 99%, Molecular Biology Grade
Potassium chloride, 99%, Molecular Biology Grade
Potassium chloride, 99%, Molecular Biology Grade
Thermo Scientific Chemicals

Potassium chloride, 99%, Molecular Biology Grade

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number J64189.36
also known as J64189-36
Price (USD)
147.65
Online exclusive
164.00
Save 16.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
147.65
Online exclusive
164.00
Save 16.35
Each
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder
Assay from Supplier's CofA≥98.5% (titration)
DNaseNot detected
Protease activityNot detected
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only