N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine, Thermo Scientific Chemicals
N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine, Thermo Scientific Chemicals
N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine, Thermo Scientific Chemicals

A cell permeable and extremely high-affinity chelator of heavy metals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
300 mg
Catalog number J64206.EXH
also known as J64206-!H
Price (USD)
292.65
Online Exclusive
325.00
Save 32.35 (10%)
Each
Quantity:
300 mg
Request bulk or custom format
Price (USD)
292.65
Online Exclusive
325.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS16858-02-9
SpecificationsSpecification SheetSpecification Sheet
Purity≥95.0%
FormPowder
A cell permeable and extremely high-affinity chelator of heavy metals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only