5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%, Thermo Scientific Chemicals
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%, Thermo Scientific Chemicals
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J64232.03
also known as J64232-03
1 g
Catalog number J64232.03
also known as J64232-03
Price (USD)
171.65
Online exclusive
191.00
Save 19.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
171.65
Online exclusive
191.00
Save 19.35
Each
Specifications
Chemical Name or Material5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine
ColorWhite to Yellow
Formula Weight657.69
MDL NumberMFCD15145247
Molecular FormulaC35H36FN5O7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only