Brilliant Blue R - FAQs
Brilliant Blue R - FAQs
View additional product information for Brilliant Blue R - FAQs (J64297.18)
1 product FAQs found
Is Brilliant Blue R (Cat. No. J64297.18) soluble in water?
Yes, Brilliant Blue R (Cat. No. J64297.18) is soluble in water. Its solubility in water is 1% with complete dissolution, to form a deep blue-black solution.