alpha-Melanocyte Stimulating Hormone amide
alpha-Melanocyte Stimulating Hormone amide
alpha-Melanocyte Stimulating Hormone amide
Thermo Scientific Chemicals

alpha-Melanocyte Stimulating Hormone amide

An endogenous melanocortin receptor agonist. Anti-inflammatory peptide; antagonizes proinflammatory mediators, including TNF-α, IL-6 and NO and induces anti-inflammatory cytokine IL-10.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J64310.LB0
also known as J64310-#0
Price (USD)
175.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
175.00
Each
Specifications
CAS581-05-5
FormatSolid
IUPAC Name(4S)-4-[(1-{[(1S)-1-{[(1S)-1-{[(1S)-1-[({[(2S)-6-amino-1-[(2S)-2-{[(1S)-1-carbamoyl-2-methylpropyl]carbamoyl}pyrrolidin-1-yl]-1-oxohexan-2-yl]carbamoyl}methyl)carbamoyl]-2-(1H-indol-3-yl)ethyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-2-phenylethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl)carbamoyl]-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-acetamido-3-hydroxypropanamido]-3-(4-hydroxyphenyl)propanamido]-3-hydroxypropanamido]-4-(methylsulfanyl)butanamido]butanoic acid
InChI KeyWHNFPRLDDSXQCL-PDEGJNOBSA-N
MDL NumberMFCD00133062
View more
alpha-Melanocyte Stimulating Hormone amide is an endogenous melanocortin receptor agonist (Ki values are 0.12, 31, 660 and 5700 nM for MC1, MC3, MC4 and MC5 receptors respectively). Anti-inflammatory peptide; antagonizes proinflammatory mediators, including TNF-α, IL-6 and NO and induces anti-inflammatory cytokine IL-10. Inhibits food intake and induces penile erections following i.c.v. administration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
Store at -20°C