Cloxacillin sodium salt
Cloxacillin sodium salt
Cloxacillin sodium salt
Thermo Scientific Chemicals

Cloxacillin sodium salt

A semisynthetic antibiotic | CAS: 642-78-4 | C19H18ClN3NaO5S | 458.869 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64314.03
also known as J64314-03
Price (USD)
54.65
Online exclusive
60.40
Save 5.75
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
54.65
Online exclusive
60.40
Save 5.75
Each
Chemical Identifiers
CAS642-78-4
IUPAC Namesodium (2S,5R,6R)-6-[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC19H17ClN3NaO5S
InChI KeySCLZRKVZRBKZCR-SLINCCQESA-M
SMILES[Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Absorbance≥825 µg/mg (anhydrous basis)
Water Content3.0-5.0%
Cloxacillin sodium salt is used to treat infections caused by penicillin G-resistant staphylococci. It has been used in studies of topical antibacterial agent for burn and crush wounds. It is used as an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cloxacillin sodium salt is used to treat infections caused by penicillin G-resistant staphylococci. It has been used in studies of topical antibacterial agent for burn and crush wounds. It is used as an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

General References:

  1. Anil Khanal.; Kenichi Nakashima. Incorporation and release of cloxacillin sodium in micelles of poly(styrene-b-2-vinyl pyridine-b-ethylene oxide). Journal of Controlled Release. 2005, 108, (1), 150-160.
  2. Els Daeseleire.; Hendrik De Ruyck and Rol.; Van Renterghem. Confirmatory assay for the simultaneous detection of penicillins and cephalosporins in milk using liquid chromatography/tandem mass spectrometry. Rapid Communications in Mass Spectrometry. 2000, 14, (15), 1404-1409.