Uridine-5'-diphosphate disodium salt, 98%
Uridine-5'-diphosphate disodium salt, 98%
Uridine-5'-diphosphate disodium salt, 98%
Thermo Scientific Chemicals

Uridine-5'-diphosphate disodium salt, 98%

CAS: 27821-45-0 | C9H12N2Na2O12P2 | 448.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number J64329.03
also known as J64329-03
Price (USD)
124.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
124.00
Each
Chemical Identifiers
CAS27821-45-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥97.5%
Uridine-5'-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine-5′-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

Solubility
Slightly soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Ivar von Kugelgen.; Klaus Starke. Evidence for two separate vasoconstriction-mediating nucleotide receptors, both distinct from the P2X-receptor, in rabbit basilar artery: a receptor for pyrimidine nucleotides and a receptor for purine nucleotides. Naunyn-Schmiedeberg's Archives of Pharmacology. 1990, 341, (6), 538-546.