4-Amino-1,8-naphthalimide, 95%
4-Amino-1,8-naphthalimide, 95%
4-Amino-1,8-naphthalimide, 95%
Thermo Scientific Chemicals

4-Amino-1,8-naphthalimide, 95%

CAS: 1742-95-6 | C12H8N2O2 | 212.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number J64358.MC
also known as J64358-MC
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.90
Save 8.25 (14%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.90
Save 8.25 (14%)
Each
Chemical Identifiers
CAS1742-95-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Purity≥94.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only