7-Methyl lumazine
7-Methyl lumazine
7-Methyl lumazine
Thermo Scientific Chemicals

7-Methyl lumazine

CAS: 13401-38-2 | C7H6N4O2 | 178.15 g/mol
Have Questions?
Catalog NumberQuantity
J64389.MB
also known as J64389-MB
25 mg
Catalog number J64389.MB
also known as J64389-MB
Price (USD)
172.65
Online exclusive
192.00
Save 19.35
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
172.65
Online exclusive
192.00
Save 19.35
Each
Specifications
CAS13401-38-2
Chemical Name or Material7-Methyl lumazine
Formula Weight178.15
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
View more
A methyl-substituted imidazo[1,2-a]pyrazine derivative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A methyl-substituted imidazo[1,2-a]pyrazine derivative with anti-inflammatory, analgesic and ulcerogenic activities.
RUO – Research Use Only