2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
Thermo Scientific Chemicals

2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid

CAS: 19231-60-8 | C10H3I4NO4 | 708.755 g/mol
Have Questions?
Catalog NumberQuantity
J64414.MA
also known as J64414-MA
10 mg
Catalog number J64414.MA
also known as J64414-MA
Price (USD)
190.65
Online exclusive
212.00
Save 21.35
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
190.65
Online exclusive
212.00
Save 21.35
Each
Specifications
CAS19231-60-8
Chemical Name or Material2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
Formula Weight708.75
IUPAC Name2-(4,5,6,7-tetraiodo-1,3-dioxoisoindol-2-yl)acetic acid
InChI KeyFLSXICNYHDJIAW-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only