Ammonium sulfate, ultrapure, 99+%
Ammonium sulfate, ultrapure, 99+%
Ammonium sulfate, ultrapure, 99+%
Thermo Scientific Chemicals

Ammonium sulfate, ultrapure, 99+%

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number J64419.22
also known as J64419-22
Price (USD)
34.50
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
34.50
Each
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition≤0.005%
Impurity contentPhosphate: ≤5 ppm
Chloride content≤5 ppm
Impurity contentNitrate: ≤0.001%
pH5.0-6.0
View more
Ammonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.

Solubility
Soluble freely in water, Insoluble in alcohol and acetone.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Gilad Haran.; Rivka Cohen.; Liliana K Bar.; Yechezkel Barenholz. Transmembrane ammonium sulfate gradients in liposomes produce efficient and stable entrapment of amphipathic weak bases. Biochimica et Biophysica Acta (BBA) - Biomembranes. 1993, 1151 (2), 201-215.
  2. Tage Astrup.; Sten Mullertz. The fibrin plate method for estimating fibrinolytic activity. Archives of Biochemistry and Biophysics. 1952, 40 (2), 346-351.