Tricine, Electrophoresis Grade, 99%
Tricine, Electrophoresis Grade, 99%
Tricine, Electrophoresis Grade, 99%
Thermo Scientific Chemicals

Tricine, Electrophoresis Grade, 99%

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number J64437.36
also known as J64437-36
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS5704-04-1
IUPAC Name2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}acetic acid
Molecular FormulaC6H13NO5
InChI KeySEQKRHFRPICQDD-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only