Sodium 4-aminosalicylate dihydrate, 98%
Sodium 4-aminosalicylate dihydrate, 98%
Sodium 4-aminosalicylate dihydrate, 98%
Thermo Scientific Chemicals

Sodium 4-aminosalicylate dihydrate, 98%

CAS: 6018-19-5 | C7H10NNaO5 | 211.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number J64449.22
also known as J64449-22
Price (USD)
67.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
67.10
Each
Chemical Identifiers
CAS6018-19-5
IUPAC Namesodium 4-amino-2-hydroxybenzoate dihydrate
Molecular FormulaC7H10NNaO5
InChI KeyGMUQJDAYXZXBOT-UHFFFAOYSA-M
SMILESO.O.[Na+].NC1=CC=C(C([O-])=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only