Azure C
Azure C
Azure C
Thermo Scientific Chemicals

Azure C

CAS: 531-57-7 | C13H12ClN3S | 277.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J64515.03
also known as J64515-03
Price (USD)
20.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
20.70
Each
Specifications
Beilstein4060661
CAS531-57-7
Chemical Name or MaterialAzure C
EINECS Number208-512-8
Formula Weight277.78
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only