G3335
G3335
G3335
G3335
Thermo Scientific Chemicals

G3335

CAS: 36099-95-3 | C16H19N3O5, C16H19N3O5 | 333.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
Catalog number J64528.MF
also known as J64528-MF
Price (USD)
128.00
Each
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)
128.00
Each
Add to cart
Specifications
CAS36099-95-3
DescriptionWhite
For Use With (Application)A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
FormSolid
Formula Weight333.34g/mol
View more
A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
RUO – Research Use Only
Contents & Storage
2°C to 8°C