Necrostatin-1, Inactive Control
Thermo Scientific Chemicals

Necrostatin-1, Inactive Control

 
Have Questions?
Catalog NumberQuantity
J64646.LB0
also known as J64646-#0
5 mg
Catalog number J64646.LB0
also known as J64646-#0
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS64419-92-7
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay (HPLC)95.0% min.
Appearance (Color)Pale yellow
A cell-permeable analog of Necrostatin-1 that is devoid of anti-necroptotic properties and is suitable as an inactive control

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only