Choline bitartrate, 98+%
Choline bitartrate, 98+%
Choline bitartrate, 98+%
Thermo Scientific Chemicals

Choline bitartrate, 98+%

CAS: 87-67-2 | C9H19NO7 | 253.251 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number J64657.30
also known as J64657-30
Price (USD)
51.10
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
51.10
Each
Chemical Identifiers
CAS87-67-2
IUPAC Name(2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular FormulaC9H19NO7
InChI KeyQWJSAWXRUVVRLH-LREBCSMRSA-M
SMILESC[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH3-4
Assay from Supplier's CofA≥98.0%
Water Content≤1.0%
FormPowder or crystals or crystalline powder
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. P Limtrakula.; S Lipigorngoson.; O Namwong.; A Apisariyakul.; F.W Dunn. Inhibitory effect of dietary curcumin on skin carcinogenesis in mice. Biochemistry. 2003, 42 (15), 4544-4551.
  2. Spector SA.; Jackman MR.; Sabounjian LA.; Sakkas C.; Landers DM.; Willis WT. Effect of choline supplementation on fatigue in trained cyclists. Bulletin of the Chemical Society of Japan. 1995, 27 (5), 668-673.