This product is animal origin and can only be shipped within the United States
Albumin, Bovine, Fraction V, 35% Solution, Reagent Grade, Fatty Acid Free, with Azide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Albumin, Bovine, Fraction V, 35% Solution, Reagent Grade, Fatty Acid Free, with Azide, Thermo Scientific Chemicals

For research use
Have Questions?
Catalog NumberQuantity
J64728.AE
also known as J64728-AE
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64728.AE
also known as J64728-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Concentration35% w/v
EINECS Number232-936-2
FormulationAqueous solution with sodium azide
Product TypeAlbumin, Bovine, Fraction V, 35% soln., Reagent grade, Fatty Acid Free, with azide
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only