N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%
Thermo Scientific Chemicals

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%

CAS: 944251-24-5 | C15H24N4O8 | 388.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J64834.MB
also known as J64834-MB
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS944251-24-5
IUPAC Name2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
Molecular FormulaC15H24N4O8
InChI KeyOIGKWPIMJCPGGD-UHFFFAOYSA-N
SMILES[N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay≥90.0%
Appearance (Color)Pale yellow to yellow
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

Solubility
Soluble in dimethyl sulfoxide and acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only