2'-O-Methyluridine, 99%
2'-O-Methyluridine, 99%
2'-O-Methyluridine, 99%
Thermo Scientific Chemicals

2'-O-Methyluridine, 99%

CAS: 2140-76-3 | C10H14N2O6 | 258.23 g/mol
Have Questions?
Catalog NumberQuantity
J64897.03
also known as J64897-03
1 g
Catalog number J64897.03
also known as J64897-03
Price (USD)
41.20
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.20
Each
Specifications
CAS2140-76-3
ChEBICHEBI:19227
Chemical Name or Material2'-O-Methyluridine
ColorWhite
Formula Weight258.23
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only