N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
Thermo Scientific Chemicals

N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate

C18H27NO9
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J64902.MC
also known as J64902-MC
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Specifications
Chemical Name or MaterialN-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
ColorOrange to Yellow
Formula Weight401.41
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate is a labeling reagent with amine reactivity as the activated NHS ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate is a labeling reagent with amine reactivity as the activated NHS ester.

Solubility
Slightly soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only