2-Aminopurine, 98% - FAQs
2-Aminopurine, 98% - FAQs
View additional product information for 2-Aminopurine, 98% - FAQs (J64919.MD, J64919.ME, J64919.03)
2 product FAQs found
What is the solubility of 2-Aminopurine 98%?
We have the following information regarding solubility for 2-Aminopurin 98%:
- Soluble in a formic acid and water solution (1:1 v/v) at 50 mg/ml by heating.
- Soluble in a glacial acetic acid and PBS solution (200:1) by heating at 60 degrees C for an 150 mM solution. This solution can then be aliquoted and stored frozen. The aliquots should be thawed, heated and mixed before usage.
What is the molecular weight of 2-Aminopurine 98%?
The molecular weight is 135.13 g/mol (this product is the non-salt form, CAS 452-06-2).