This product is animal origin and can only be shipped within the United States
Avidin, from egg
Thermo Scientific Chemicals

Avidin, from egg

 
Have Questions?
Catalog NumberQuantity
J64972.MA
also known as J64972-MA
10 mg
Catalog number J64972.MA
also known as J64972-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
CAS1405-69-2
ColorWhite
EINECS Number215-783-6
For Use With (Application)Has extremely strong affinity for biotin
Product TypeAvidin
View more
Has extremely strong affinity for biotin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C