Azadibenzocyclooctyne-PEG, MW 5,000
Thermo Scientific Chemicals

Azadibenzocyclooctyne-PEG, MW 5,000

 
Have Questions?
Catalog NumberQuantity
J64977.MC
also known as J64977-MC
100 mg
Catalog number J64977.MC
also known as J64977-MC
Price (USD)
224.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
224.00
Each
Specifications
Chemical Name or MaterialAzadibenzocyclooctyne-PEG
ColorWhite to Gray
Formula Weight5kDa
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool,
RUO – Research Use Only

General References:

  1. Rodrigo F. Ortiz-Meoz.; Yifat Merbl.; Marc W. Kirschner.; Suzanne Walker. Microarray Discovery of New OGT Substrates: The Medulloblastoma Oncogene OTX2 Is O-GlcNAcylated. J. Am. Chem. Soc. 2014, 136, (13), 4845-4848.
  2. Nicole Nischan.; Christian P. R. Hackenberger. Site-specific PEGylation of Proteins: Recent Developments. J. Org. Chem. 2014, 79, (22), 10727-10733.