2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J64981.MA
also known as J64981-MA
Price (USD)
128.65
Special offer
Online exclusive
Ends: 30-Jun-2026
151.00
Save 22.35 (15%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
128.65
Special offer
Online exclusive
Ends: 30-Jun-2026
151.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS418788-90-6
SpecificationsSpecification SheetSpecification Sheet
Purity≥97.0% (HPLC)
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only