Orexin B, Human
Thermo Scientific Chemicals

Orexin B, Human

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J65110.EXD
also known as J65110-!D
Price (USD)
218.65
Online Exclusive
243.00
Save 24.35 (10%)
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
218.65
Online Exclusive
243.00
Save 24.35 (10%)
Each
Specifications
FormatLyophilized Powder
Molecular Weight (g/mol)2899.33
Product TypeOrexin B, human
Purification MethodHPLC
Quantity0.5 mg
View more

Orexin is a neuropeptide that modulates the energy and metabolism balance. The effects of orexin are mediated by one of the two G protein-coupled receptors, orexin receptor 1 or 2. OXRs are expressed in oxinergic projection areas of the brain and regulate food intake by reducing systemic and brain signals that drive over-eating. Synthetic peptide corresponding to the C-terminus of human orexin receptor 1, conjugated to KLH. The immunizing peptide has 70% homology with the rat and mouse gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C