Stem-Cell Factor/c-Kit Inhibitor
Thermo Scientific Chemicals
Stem-Cell Factor/c-Kit Inhibitor
Have Questions?
Catalog NumberQuantity
J65114.MA
also known as J65114-MA
10mg
Catalog number J65114.MA
also known as J65114-MA
Price (USD)
133.65
Online exclusive
148.00
Save 14.35
Each
Quantity:
10mg
Request bulk or custom format
Price (USD)
133.65
Online exclusive
148.00
Save 14.35
Each
Specifications
Molecular Weight (g/mol)355.50
PackagingPlastic bottle
Quantity10mg
SensitivityLight sensitive
FormSolid
View more
Inhibits in vitro phosphorylation of c-Kit. Also inhibits SCF (stem cell factor)-induced c-kit phosphorylation in 501mel human melanoma cells