8-Anilinonaphthalene-1-sulfonic acid, 95%
8-Anilinonaphthalene-1-sulfonic acid, 95%
8-Anilinonaphthalene-1-sulfonic acid, 95%
Thermo Scientific Chemicals

8-Anilinonaphthalene-1-sulfonic acid, 95%

Fluorescent probe for proteins on polyacrylamide gels | CAS: 82-76-8 | C16H13NO3S | 299.344 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number J65332.14
also known as J65332-14
Price (USD)
116.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
116.00
Each
Chemical Identifiers
CAS82-76-8
IUPAC Name8-(phenylamino)naphthalene-1-sulfonic acid
Molecular FormulaC16H13NO3S
InChI KeyFWEOQOXTVHGIFQ-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale grey to black or brown
FormCrystals or powder or crystalline powder
Assay by NMR≥94.0% (Proton-NMR)
Fluorescent probe for proteins on polyacrylamide gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only