Cafestol
Cafestol
Cafestol
Thermo Scientific Chemicals

Cafestol

Natural extract from coffee beans which induces glutathione S-transferase | CAS: 469-83-0 | C20H28O3 | 316.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
Catalog number J65355.MC
also known as J65355-MC
Price (USD)
543.00
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
543.00
Each
Add to cart
Chemical Identifiers
CAS469-83-0
IUPAC Name(1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
Molecular FormulaC20H28O3
InChI KeyDNJVYWXIDISQRD-HWUKTEKMSA-N
SMILESC[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off white
FormPowder
Assay (HPLC)≥ 98.0% (UHPLC)
Assay (special)Identification: Conforms
Natural extract from coffee beans which induces glutathione S-transferase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only