8-Bromo-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals
8-Bromo-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals
8-Bromo-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals
8-Bromo-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Bromo-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J65357.03
also known as J65357-03
1 g
Catalog number J65357.03
also known as J65357-03
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Add to cart
Specifications
CAS14985-44-5
Chemical Name or Material8-Bromo-2'-deoxyadenosine
ColorWhite to Yellow
Formula Weight330.14
Health Hazard 1H302-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only